missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Importin 13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521884
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521884 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521884 Supplier Invitrogen™ Supplier No. PA521884
Only null left

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa, Importin 13 tansfected 293T cell. Predicted reactivity: Mouse (100%), Rat (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the importin-beta family of nuclear transport proteins. The encoded protein mediates the import of specific cargo proteins from the cytoplasm to the nucleus and is dependent on the Ras-related nuclear protein-GTPase system. The encoded protein is also involved in nuclear export of the eukaryotic translation initiation factor 1A.

Specifications

Antigen Importin 13
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.02 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene IPO13
Gene Accession No. O94829, Q9JM04
Gene Alias Imp13; importin 13; importin-13; Ipo13; Kap13; karyopherin 13; karyopherin-13; KIAA0724; late gestation lung 2; late gestation lung 2 protein; LGL2; Ran binding protein 13; ran-binding protein 13; RanBP13; RP11-7O11.4
Gene Symbols IPO13
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Importin 13. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116458, 9670
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less