Learn More
StatLab™ Immunocal Decalcifier
Ideal for decalcifying bone for IHC.
$75.21 - $175.39
Specifications
Solution Type Decalcifier
View More Specs
Alert:
Harmful if swallowed, in contact with skin, or inhaled.
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
STL141432
View Documents Promotion Details
StatLab™
STL141432
32 oz.
Each for $75.21
Only null left
STL14141
View Documents Promotion Details
StatLab™
STL14141
1 gal.
Each for $175.39
Case of 4 Each for $701.56
Only null left
Description
Description
  • A very mild formic acid decalcifier formulated from semiconductor grade formic acid and highly purified reagent grade water
  • Will not destroy antigen sites during decalcification
  • Can safely be used to demonstrate antigens in decalcified bone
Specifications
Specifications
Decalcifier