Learn More
StatLab™ Immunocal Decalcifier
Catalog No. STL141432
Change view
Click to view available options
Quantity:
32 oz.
1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STL141432 32 oz.
STL14141 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STL141432 Supplier StatLab™ Supplier No. STL141432
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Harmful if swallowed, in contact with skin, or inhaled.
Ideal for decalcifying bone for IHC.
  • A very mild formic acid decalcifier formulated from semiconductor grade formic acid and highly purified reagent grade water
  • Will not destroy antigen sites during decalcification
  • Can safely be used to demonstrate antigens in decalcified bone
Specifications
Quantity 32 oz.
Solution Type Decalcifier