missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Immobilon™ UltraPlus Western HRP Substrate

Catalog No. WBULP20ML
Change view
Click to view available options
Quantity:
10 mL total
100 mL total
20 mL total
Content And Storage:
10 mL Enhancer, 10 mL Peroxide
5 mL Enhancer, 5 mL Peroxide
50 mL Enhancer, 50 mL Peroxide
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Content And Storage
WBULP20ML 20 mL total 10 mL Enhancer, 10 mL Peroxide
WBULP10ML 10 mL total 5 mL Enhancer, 5 mL Peroxide
WBULP100ML 100 mL total 50 mL Enhancer, 50 mL Peroxide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. WBULP20ML Supplier MilliporeSigma™ Supplier No. WBULP20ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Immobilon™ UltraPlus delivers maximum sensitivity for Western blot immunodetection, and is ideal for detection of low-abundance targets.

  • A non-isotopic, luminol-based chemiluminescence substrate for Horseradish Peroxidase (HRP), designed for the chemiluminescent detection of immobilized proteins conjugated with horseradish peroxidase (HRP).
  • It is compatible with both PVDF and nitrocellulose blotting membranes, as well as commonly used buffers and blocking reagents.
  • For research use only
  • Storage Class Code: 10 - Combustible liquids

Specifications

For Use With (Application) Dot Blot, Immunoblotting, Western Blot
Quantity 20 mL total
Content And Storage 10 mL Enhancer, 10 mL Peroxide
Product Line Immobilon

Unless otherwise stated, this product is intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.