Learn More
MilliporeSigma™ Immobilon Signal Enhancer for Immunodetection
Catalog No. WBSH0500
Change view
Click to view available options
Quantity:
500 mL
Catalog No. Quantity
WBSH0500 500 mL
1 options
Catalog No. WBSH0500 Supplier MilliporeSigma™ Supplier No. WBSH0500
Includes
Immobilon Signal Enhancer for Immunodetection 500ML
MilliporeSigma™ Immobilon™ Signal Enhancer for Immunodetection combines signal amplification and blocking in one ready-to-use reagent. Amplifies low signal intensity Western blots without amplifying the noise
  • Immobilon™ Signal Enhancer is designed for use in Western blotting applications when low signal intensity is observed. It combines signal amplification and blocking in one ready-to-use reagent.
  • Enhancement of the antibody-antigen reaction can produce a 2 to 10-fold increase in signal intensity over traditionally used reagents. The reagent is formulated for use in both membrane blocking and antibody dilution steps and can be used for traditional immunodetection or with the SNAP i.d.™ Protein Detection System.
  • Save Primary Antibody: When the signal is strong, use Immobilon™ Signal Enhancer to reduce the amount of valuable primary antibody required
  • Immobilon™ Signal Enhancer is compatible with PVDF as well as nitrocellulose blotting membranes and can be used with chromogenic, chemiluminescent, and fluorescent detection methods.
  • In applications where sufficient signal is observed, but the primary antibody availability is limited, Immobilon™ Signal Enhancer can be used to reduce the amount of valuable primary antibody required
Certifications
request
Specifications
For Use With (Application) Western Blotting, immunodetection
Quantity 500 mL
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.