Learn More
MilliporeSigma™ Immobilon ECL Ultra Western HRP Substrate
MilliporeSigma™ Immobilon ECL Ultra Western HRP Substrate excels in all Western and dot/slot/spot blotting applications. Compatible with both PVDF and nitrocellulose membranes, as well as all commonly used buffers and blocking reagents.
Supplier:  MilliporeSigma™ WBULS0100
 View more versions of this product
Catalog No. WBULS0100
Only null left
Explore available promotions
Includes: Immobilon ECL Ultra Western HRP Substrate 100ML: Solution A- 50 mL; Solution B- 50 mL
Description
Description
Features:
  • Immobilon™ ECL Ultra Western HRP Substrate is an ultra-sensitive reagent with long signal duration for the detection of horseradish peroxidase (HRP)-labeled antibodies on western blots.
  • The intense signal created permits sensitive detection at the mid-to-low femtogram range using x-ray film or digital imaging systems.
  • Immobilon™ ECL Ultra substrate is compatible with both PVDF and nitrocellulose blotting membranes, as well as commonly used buffers and blocking reagents.
  • Sufficient for 1000 cm²
Certifications
request
Specifications
Specifications
Western blotting, HRP
100 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.