missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Immobilon™-E Transfer Membrane
Immobilon™-E provides the a PVDF blotting membrane that wets in transfer buffer or water and eliminates the need for the alcohol pre-wet step.
Supplier: MilliporeSigma™ IEVH07804
Description
Features:
- 0.45 μm pore size is ideal for most Western blotting applications with chemiluminescence or chromogenic detection
- Saves time and reduces waste by eliminating the alcohol pre-wet step
- Higher protein retention delivers stronger signal than most nitrocellulose membranes
Specifications
| Easily wetted with water | |
| White | |
| Plain | |
| Sheet | |
| 0.45 μm |
| Western Blotting | |
| IEVH | |
| 4 Sheets | |
| PVDF | |
| 7 x 8.4 cm |