missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Immobilon™-E Transfer Membrane

Catalog No. IEVH00005
Change view
Click to view available options
Quantity:
1 Roll
10 Sheets
4 Sheets
50 Sheets
Dimensions (LxW):
1.875 m x 26.5 cm
10 m x 8.5 cm
7 x 8.4 cm
8 x 10 cm
9 x 12 cm
10 x 10 cm
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Dimensions (LxW)
IEVH00005 1 Roll 1.875 m x 26.5 cm
IEVH85R 1 Roll 10 m x 8.5 cm
IEVH07804 4 Sheets 7 x 8.4 cm
IEVH07850 50 Sheets 7 x 8.4 cm
IEVH08100 10 Sheets 8 x 10 cm
IEVH09120 10 Sheets 9 x 12 cm
IEVH10100 10 Sheets 10 x 10 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. IEVH00005 Supplier MilliporeSigma™ Supplier No. IEVH00005

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Immobilon™-E provides the a PVDF blotting membrane that wets in transfer buffer or water and eliminates the need for the alcohol pre-wet step.

Features:

  • 0.45 μm pore size is ideal for most Western blotting applications with chemiluminescence or chromogenic detection
  • Saves time and reduces waste by eliminating the alcohol pre-wet step
  • Higher protein retention delivers stronger signal than most nitrocellulose membranes

Specifications

Length (Metric) 1.875 m
Width (Metric) 27 cm
Description Easily wetted with water
For Use With (Application) Western Blotting
Color White
Code IEVH
Surface Plain
Quantity 1 Roll
Format Roll
Material PVDF
Pore Size 0.45 μm
Dimensions (LxW) 1.875 m x 26.5 cm
Show More Show Less