Learn More
Cytiva Immobiline™ DryStrip Gels
Represent a comprehensive selection of narrow-, medium- and wide-range IPG strips. Cytiva Life Sciences™ Immobiline™ DryStrip gels (IPG strips) are used for isoelectric focusing (IEF), run as the first dimension of 2-D electrophoresis or as a separate application.
$226.95 - $1,030.81
Specifications
Description Immobiline DryStrip gels (IPG strips) are used for isoelectric focusing (IEF), run as the first dimension of 2-D electrophoresis or as a separate application.
Color Colorless
Format Precast Gel
Separation Type Isoelectric Focusing
Form Dried
View More Specs
Products 24
Catalog Number Mfr. No. pH Range Gradient Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. pH Range Gradient Length (Metric) Price Quantity & Availability  
45000213
View Documents Promotion Details
Cytiva
17123501
3 to 10 Non-linear 18 cm
Pack of 12 Each for $319.69
Only null left
45000362
View Documents Promotion Details
Cytiva
17600197
6 to 11 Linear 18 cm
Pack of 12 Each for $577.47
Only null left
45000361
View Documents Promotion Details
Cytiva
17600196
6 to 11 Linear 13 cm
Pack of 12 Each for $1,030.81
Only null left
45000359
View Documents Promotion Details
Cytiva
17600194
6 to 11 Linear 7 cm
Pack of 12 Each for $464.54
Only null left
45000379
View Documents Promotion Details
Cytiva
17600247
6 to 9 Linear 24 cm
Pack of 12 Each for $750.11
Only null left
45000377
View Documents Promotion Details
Cytiva
17600245
3 to 10 Non-linear 24 cm
Pack of 12 Each for $373.07
Only null left
45000402
View Documents Promotion Details
Cytiva
17600373
3 to 11 Non-linear 7 cm
Pack of 12 Each for $230.84
Only null left
45000404
View Documents Promotion Details
Cytiva
17600375
3 to 11 Non-linear 13 cm
Pack of 12 Each for $321.71
Only null left
45000406
View Documents Promotion Details
Cytiva
17600377
3 to 11 Non-linear 24 cm
Pack of 12 Each for $379.46
Only null left
45000211
View Documents Promotion Details
Cytiva
17123301
4 to 7 Linear 18 cm
Pack of 12 Each for $319.69
Only null left
45000212
View Documents Promotion Details
Cytiva
17123401
3 to 10 Linear 18 cm
Pack of 12 Each for $284.32
Only null left
45000347
View Documents Promotion Details
Cytiva
17600111
3 to 10 Linear 7 cm
Pack of 12 Each for $230.84
Only null left
45000348
View Documents Promotion Details
Cytiva
17600112
3 to 10 Non-linear 7 cm
Pack of 12 Each for $226.95
Only null left
45000346
View Documents Promotion Details
Cytiva
17600110
4 to 7 Linear 7 cm
Pack of 12 Each for $226.95
Only null left
45000349
View Documents Promotion Details
Cytiva
17600113
4 to 7 Linear 13 cm
Pack of 12 Each for $319.69
Only null left
45000350
View Documents Promotion Details
Cytiva
17600114
3 to 10 Linear 13 cm
Pack of 12 Each for $319.69
Only null left
45000351
View Documents Promotion Details
Cytiva
17600115
3 to 10 Non-linear 13 cm
Pack of 12 Each for $319.69
Only null left
45000360
View Documents Promotion Details
Cytiva
17600195
6 to 11 Linear 11 cm
Pack of 12 Each for $889.00
Only null left
45000378
View Documents Promotion Details
Cytiva
17600246
4 to 7 Linear 24 cm
Pack of 12 Each for $379.46
Only null left
45000376
View Documents Promotion Details
Cytiva
17600244
3 to 10 Linear 24 cm
Pack of 12 Each for $373.07
Only null left
45000405
View Documents Promotion Details
Cytiva
17600376
3 to 11 Non-linear 18 cm
Pack of 12 Each for $319.69
Only null left
45000403
View Documents Promotion Details
Cytiva
17600374
3 to 11 Non-linear 11 cm
Pack of 12 Each for $252.47
Only null left
45000428
View Documents Promotion Details
Cytiva
18101660
4 to 7 Linear 11 cm
Pack of 12 Each for $226.95
Only null left
45000429
View Documents Promotion Details
Cytiva
18101661
3 to 10 Linear 11 cm
Pack of 12 Each for $252.47
Only null left
Description
Description
Choose from a comprehensive range of overlapping Immobiline DryStrip gels that cover narrow (1 pH unit), medium (3 to 5 pH units), and broad (8 pH units) pH ranges. The pH gradients at the extreme ends of the pH scale are nonlinear to distribute the proteins evenly over the gel length to obtain maximum resolution.
Five narrow pH range (1 pH unit), 24-cm IPG strips deliver high resolution and protein loading capacity, which results in well separated spots with high protein content after the second-dimension separation
  • Select the broad pH 3 to 11 Non-Linear (NL) gradient Immobiline DryStrip gel for fast and efficient screening to gain a broad overview of total protein disribution
  • For more detailed studies or when using greater protein loads, choose from four new overlapping strips covering the pH 3 to 11 range
  • Higher resolution can be obtained using narrow-range (1 pH unit) Immobiline DryStrip gels
  • Use matching IPG Buffers for optimized running conditions
  • Use IPG strips directly after reswelling without further manipulation
  • Bar coded strips for convenient sample and batch tracking
  • Immobiline DryStrip gels are rehydrated with a matching IPG Buffer prior to use
  • Samples can be included directly in the rehydration solution for efficient loading of dilute samples or for loading larger amounts of protein
  • Samples can also be loaded after rehydration
Specifications
Specifications
Immobiline DryStrip gels (IPG strips) are used for isoelectric focusing (IEF), run as the first dimension of 2-D electrophoresis or as a separate application.
Precast Gel
Dried
−20°C
0.05 cm
0.3 cm
Colorless
Isoelectric Focusing
Industrial applications: analytical chemistry, research
0.19 in.
0.118 in.
Precast Gel
Videos
Safety and Handling
Safety and Handling
ShelfLife : 18 Months