Learn More
Thermo Scientific™ Imject™ Maleimide-Activated Blue Carrier™ Protein
Catalog No. PI77664
Change view
Click to view available options
Quantity:
2 mg
10 mg
10 x 10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI77664 10 mg
PI77661 2 mg
PI77662 10 x 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PI77664 Supplier Thermo Scientific™ Supplier No. 77664
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Make hapten-carrier immunogens comprising cysteine-containing peptides and other thiolated antigens with sulfhydryl-reactive immunogenic hemocyanin (CCH).
Thermo Scientific™ Maleimide-Activated Blue Carrier Protein is sulfhydryl-reactive immunogenic hemocyanin (CCH) for hapten-carrier conjugation with cysteine-containing peptides and other thiolated antigens.
Blue Carrier Protein is a large homocyanin that is highly immunogenic like keyhole limpet hemocyanin (KLH) yet is more soluble. The carrier is rendered sulfhydryl-reactive by chemical modification with Sulfo-SMCC crosslinker, and the resulting maleimide-activated protein is then stabilized and lyophilized in phosphate buffer. Maleimide groups form covalent crosslinks with sulfhydryl (-SH) moieties on cysteine residues of peptides and other thiol-containing haptens. Numerous antigen molecules can be conjugated to each Blue Carrier Protein molecule to make them highly immunogenic and increase the likelihood of eliciting immunized animals to produce high-titer antisera to the epitopes of interest.
Highlights:
  • Optimized activation – specifications require that each lot is activated with more than 400 moles of sulhydryl-reactive maleimide per mole of carrier, ensuring consistently high levels of conjugatio
  • Sulfhydryl-reactive – engineered for conjugating cysteine-containing peptide antigens to make effective immunogens for high-titer antibody production
  • Simple, rapid procedure – complete the hapten-carrier conjugation reaction in two hours to yield sufficient immunogen for several injections and booster immunizations of host animals
  • High-quality carrier protein – Blue Carrier Protein is ultrapure Concholepas concholepas hemocyanin (CCH) prepared from molluscan hemolymph
  • Highly immunogenic and soluble – CCH has a high molecular mass that elicits a stronger immune response than BSA or ovalbumin
Specifications
Product Line Blue Carrier, Imject
Product Type Imject Maleimide-Activated Blue Carrier Protein
Content And Storage Upon receipt store at -20°C. Product is shipped on ice.
Quantity 10 mg
For Research Use Only. Not for use in diagnostic procedures.