missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Imject™ Freund's Complete Adjuvant (FCA)

Catalog No. pi77140
Change view
Click to view available options
Quantity:
5 x 10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI77140 5 x 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI77140 Supplier Thermo Scientific™ Supplier No. 77140
Only null left

Stimulate a nonspecific immune response using complete (CFA) or incomplete (IFA) formulations of the popular Freund's adjuvant; offered in 10mL ampules.

Thermo Scientific™ Imject Freund's Adjuvants are 10mL ampules of complete (FCA) and incomplete (FIA) formulations of a popular nonspecific stimulator of the immune response for use in antibody production immunization protocols.

The water-and-oil emulsions (with and without inactivated Mycobacterium) greatly enhance immune responses to immunogens when mixed and injected with the prepared antigen. Typically, FCA is used for initial injections and FIA is used for booster injections. The ampules comprise conveniently sized aliquots that provide long shelf life.

Highlights:

  • Water-in-oil emulsion with or without killed Mycobacterium
  • Enhance immune response and antibody production for injected immunogens
  • Useful for immunization of mice, rabbits and other mammalian hosts
  • Requires emulsification with prepared immunogens

Freund's Complete Adjuvant (FCA), Part No. 77140:

  • Water-in-oil emulsion containing killed Mycobacterium
  • Greatly enhances immune response to immunogen
  • Typically used for initial injections
  • Disadvantages: difficult to mix with immunogen and can cause tissue necrosis at injection site

Freund's Incomplete Adjuvant (FIA), Part No. 77145:

  • Water-in-oil emulsion without Mycobacterium
  • Enhances immune response in booster injections
  • Typically used for subsequent boosts after initial injection with FCA
  • Disadvantage: difficult to mix with immunogen

Specifications

Product Line Imject
Quantity 5 x 10 mL
Description Imject Freund's Complete Adjuvant (FCA)
Content And Storage Upon receipt store at 4°C

For Research Use Only. Not for use in diagnostic procedures.