missing translation for 'onlineSavingsMsg'
Learn More

Imipramine hydrochloride

Catalog No. AAJ6372314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6372314 25 g
AAJ6372306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6372314 Supplier Thermo Scientific Chemicals Supplier No. J6372314

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A serotonin uptake inhibitor | CAS: 113-52-0 | C19H25ClN2 | 316.873 g/mol

Imipramine hydrochloride is used as a serotonin uptake inhibitor. It mainly used in the treatment of major depression and enuresis (inability to control urination). It has also been evaluated for use in panic disorder.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Imipramine hydrochloride is used as a serotonin uptake inhibitor. It mainly used in the treatment of major depression and enuresis (inability to control urination). It has also been evaluated for use in panic disorder.

Solubility
Soluble in water

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 113-52-0
Molecular Formula C19H25ClN2
Molecular Weight (g/mol) 316.873
MDL Number MFCD00012669
InChI Key XZZXIYZZBJDEEP-UHFFFAOYSA-N
Synonym imipramine hydrochloride, imipramine hcl, tofranil, chimoreptin, chrytemin, deprinol, feinalmin, imilanyle, persamine, pertofram
PubChem CID 8228
ChEBI CHEBI:5882
IUPAC Name 3-(5,6-dihydrobenzo[b][1]benzazepin-11-yl)-N,N-dimethylpropan-1-amine;hydrochloride
SMILES CN(C)CCCN1C2=CC=CC=C2CCC3=CC=CC=C31.Cl

Specifications

Melting Point 174°C to 175°C
Color White
Quantity 25 g
Merck Index 14,4920
Solubility Information Soluble in water
Formula Weight 316.87
Physical Form Crystalline Powder
Chemical Name or Material Imipramine hydrochloride
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 204-030-7
RTECSNumber HO1925000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only