accessibility menu, dialog, popup

UserName

CAS RN 113-52-0

IUPAC Name: hydrogen (3-{2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-yl}propyl)dimethylamine chloride
Synonyms: hydrogen imipramine chloride
Molecular Weight (g/mol): 316.87
Molecular Formula: C19H25ClN2
InChi Key: XZZXIYZZBJDEEP-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN(C)CCCN1C2=CC=CC=C2CCC2=CC=CC=C12
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)