missing translation for 'onlineSavingsMsg'
Learn More

Imipramine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7131689
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
I09711G 1 g
I09715G 5 g
2 options

Catalog No. I09711G

Supplier: TCI America I09711G

Only null left

Chemical Identifiers

CAS 113-52-0
Molecular Formula C19H25ClN2
Molecular Weight (g/mol) 316.873
MDL Number MFCD00012669
InChI Key XZZXIYZZBJDEEP-UHFFFAOYSA-N
Synonym imipramine hydrochloride, imipramine hcl, tofranil, chimoreptin, chrytemin, deprinol, feinalmin, imilanyle, persamine, pertofram
PubChem CID 8228
ChEBI CHEBI:5882
IUPAC Name 3-(5,6-dihydrobenzo[b][1]benzazepin-11-yl)-N,N-dimethylpropan-1-amine;hydrochloride
SMILES CN(C)CCCN1C2=CC=CC=C2CCC3=CC=CC=C31.Cl

Specifications

Melting Point 173°C
Color White-Yellow
Quantity 1 g
Formula Weight 316.87
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Imipramine Hydrochloride
RTECSNumber HO1925000
TSCA Yes