missing translation for 'onlineSavingsMsg'
Learn More

Imipramine Hydrochloride 98.0+%, TCI America™

Catalog No. I09715G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09715G 5 g
I09711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09715G Supplier TCI America Supplier No. I09715G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 113-52-0
Molecular Formula C19H25ClN2
Molecular Weight (g/mol) 316.873
MDL Number MFCD00012669
InChI Key XZZXIYZZBJDEEP-UHFFFAOYSA-N
Synonym imipramine hydrochloride, imipramine hcl, tofranil, chimoreptin, chrytemin, deprinol, feinalmin, imilanyle, persamine, pertofram
PubChem CID 8228
ChEBI CHEBI:5882
IUPAC Name 3-(5,6-dihydrobenzo[b][1]benzazepin-11-yl)-N,N-dimethylpropan-1-amine;hydrochloride
SMILES CN(C)CCCN1C2=CC=CC=C2CCC3=CC=CC=C31.Cl

Specifications

Melting Point 173°C
Color White-Yellow
Quantity 5 g
Formula Weight 316.87
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Imipramine Hydrochloride
RTECSNumber HO1925000
TSCA Yes