missing translation for 'onlineSavingsMsg'
Learn More

Imipramine Hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012634
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012634 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012634 Supplier Merck Emd Millipore Supplier No. PHR17971G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Imipramine Hydrochloride is typically utilized for the treatment of depressive disorders due to its efficiency in uplifting the patients′ mood by interfering in the reuptake of norepinephrine or serotonin. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 113-52-0
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C19H24N2 · HCl
MDL Number MFCD00012669
Synonym Imipramine hydrochloride; 10,11-Dihydro-N,N-dimethyl-5 H-dibenz[b,f]azepine-5-propanamine hydrochloride; 5-[3-(Dimethylamino)propyl]-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride
Type Certified Reference Material
Quantity 1 g
Formula Weight 316.87
Grade Certified Reference Material