missing translation for 'onlineSavingsMsg'
Learn More

Iminodiacetonitrile, 95%

Catalog No. AAB2366022
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2366022 100 g
AAB2366036 500 g
AAB236600E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2366022 Supplier Thermo Scientific Chemicals Supplier No. B2366022

CAS: 628-87-5 | C4H5N3 | 95.11 g/mol

Iminodiacetonitrile is used as a chemical intermediate. It is also used in the preparation of iminodiacetic acid. Further, it is used to study the interference of nitriles in cyanide determination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iminodiacetonitrile is used as a chemical intermediate. It is also used in the preparation of iminodiacetic acid. Further, it is used to study the interference of nitriles in cyanide determination.

Solubility
Soluble in water, acetone and hot methanol.

Notes
Incompatible with strong acids and strong oxidizing agents.

Chemical Identifiers

CAS 628-87-5
Molecular Formula C4H5N3
Molecular Weight (g/mol) 95.11
MDL Number MFCD00001887
InChI Key BSRDNMMLQYNQQD-UHFFFAOYSA-N
Synonym iminodiacetonitrile, 2,2'-azanediyldiacetonitrile, acetonitrile, 2,2'-iminobis, 1,1'-imidodiacetonitrile, acetonitrile, iminodi, 2,2'-iminodiacetonitrile, 2,2'-iminobisacetonitrile, bis cyanomethyl amine, di cianometil ammina, 2-cyanomethyl amino acetonitrile
PubChem CID 69413
ChEBI CHEBI:71189
IUPAC Name 2-(cyanomethylamino)acetonitrile
SMILES N#CCNCC#N

Specifications

Melting Point 75°C to 79°C
Quantity 100 g
Beilstein 773803
Solubility Information Soluble in water,acetone and hot methanol.
Formula Weight 95.11
Percent Purity 95%
Chemical Name or Material Iminodiacetonitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
EINECSNumber 211-058-3
RTECSNumber AM0650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only