missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Iminodiacetic acid, 98+%

Catalog No. AC204970250
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC204970250 25 g
AC204975000 500 g
AC204970025 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC204970250 Supplier Thermo Scientific Chemicals Supplier No. 204970250
Only null left

CAS: 142-73-4 | C4H7NO4 | 133.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 142-73-4
Molecular Formula C4H7NO4
Molecular Weight (g/mol) 133.1
MDL Number MFCD00004280
InChI Key NBZBKCUXIYYUSX-UHFFFAOYSA-N
Synonym iminodiacetic acid, 2,2'-iminodiacetic acid, glycine, n-carboxymethyl, aminodiacetic acid, diglycocoll, diglykokoll, diglycin, iminodiethanoic acid, n-carboxymethyl glycine, 2,2'-azanediyldiacetic acid
PubChem CID 8897
ChEBI CHEBI:24786
IUPAC Name 2-(carboxymethylamino)acetic acid
SMILES C(C(=O)O)NCC(=O)O

Specifications

Melting Point 243°C
Color White or Yellow
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Plastic bottle
Linear Formula HN(CH2CO2H)2
Quantity 25 g
Beilstein 04, 365
Merck Index 15, 4959
Solubility Information Solubility in water: 2.43g/100 mL water (5°C). Other solubilities: 2.9g/100 mL water (20°C),6.3g/100 mL water (40°C),12.8g/100 mL water (60°C),23.0g/100 mL water (80°C),32.5g/100 mL water (100°C)
Formula Weight 133.1
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material Iminodiacetic acid, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 205-555-4
RTECSNumber AI2975000
TSCA TSCA

RUO – Research Use Only