accessibility menu, dialog, popup

UserName

HOONHOHO

CAS RN 142-73-4

IUPAC Name: 2-[(carboxymethyl)amino]acetic acid
Synonyms: iminodiacetic acid IDA diglycocoll iminodiethanoic acid diglycine
Molecular Weight (g/mol): 133.1
Molecular Formula: C4H7NO4
InChi Key: NBZBKCUXIYYUSX-UHFFFAOYSA-N
SMILES: OC(=O)CNCC(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (5)