missing translation for 'onlineSavingsMsg'
Learn More

Imidazole Slightly off-white, crystalline powder MP Biomedicals

Catalog No. icn10203301
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10203301 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10203301 Supplier MP Biomedicals Inc Supplier No. 0210203301
Only null left

Chemical Identifiers

CAS 288-32-4
Molecular Formula C3H4N2
Molecular Weight (g/mol) 68.08
MDL Number MFCD00005183
InChI Key RAXXELZNTBOGNW-UHFFFAOYSA-N
Synonym imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole
PubChem CID 795
ChEBI CHEBI:16069
IUPAC Name 1H-imidazole
SMILES N1C=CN=C1

Specifications

Color White
Density 500 - 600 kg/m3 (bulk density)
Quantity 1 kg
Formula Weight 68.1
Physical Form Crystalline Powder
Chemical Name or Material Imidazole