missing translation for 'onlineSavingsMsg'
Learn More

Imidazole, Reagent, 99%, Spectrum™ Chemical

Catalog No. p-7157633
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603420 25 g Poly Bottle
18603421 100 g Poly Bottle
18603422 500 g Poly Bottle
18603423 1 kg Poly Pail
18603424 5 kg Poly Pail
18603425 12 kg Poly Pail
18603426 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18603420 Supplier Spectrum Chemical Mfg Cor Supplier No. IM10525GM
Only null left

IM105, 288-32-4, C3H4N2

Spectrum™ Chemical Imidazole, Reagent is a solid that has no color and is water soluble. It is classified as an diazole and an alkaloid and is an aromatic heterocycle. It belongs to the class of antifungals known as azole and is also used as a corrosion inhibitor on certain metals, notably copper. Imidazole is incorporated into many biological molecules, primarily histidine. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Chemical Identifiers

CAS 288-32-4
Molecular Formula C3H4N2
Molecular Weight (g/mol) 68.08
MDL Number MFCD00005183
InChI Key RAXXELZNTBOGNW-UHFFFAOYSA-N
IUPAC Name 1H-imidazole
SMILES N1C=CN=C1

Specifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 25 g
UN Number UN3263
Percent Purity 99%
Grade Reagent