missing translation for 'onlineSavingsMsg'
Learn More

Imidazole, Molecular Biology Grade, 99+%, Thermo Scientific Chemicals

$427.21 - $427.21

Chemical Identifiers

CAS 288-32-4
Molecular Formula C3H4N2
Molecular Weight (g/mol) 68.08
MDL Number MFCD00005183
InChI Key RAXXELZNTBOGNW-UHFFFAOYSA-N
Synonym imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole
PubChem CID 795
ChEBI CHEBI:16069
SMILES N1C=CN=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ671326Y
View Documents
Thermo Scientific Chemicals
J671326Y
2 x 25 g N/A
Only null left
 
Description

Description

Suitable for molecular biology applications

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

288-32-4
68.08
RAXXELZNTBOGNW-UHFFFAOYSA-N
795
N1C=CN=C1
C3H4N2
MFCD00005183
imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole
CHEBI:16069

Specifications

288-32-4
White
256°C
MFCD00005183
UN2923
144,912
RAXXELZNTBOGNW-UHFFFAOYSA-N
1 H-imidazole
795
68.08
Molecular Biology
Imidazole
90°C
1.03 g/mL
C3H4N2
2 x 25 g
103853
imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole
N1C=CN=C1
68.08
CHEBI:16069
≥99%
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

RTECSNumber : NI3325000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only