missing translation for 'onlineSavingsMsg'
Learn More

Imidazole Hydrobromide (Low water content) 98.0+%, TCI America™

Catalog No. I10061G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I10061G 1 g
I10065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I10061G Supplier TCI America Supplier No. I10061G

Chemical Identifiers

CAS 101023-55-6
Molecular Formula C3H5BrN2
Molecular Weight (g/mol) 148.99
MDL Number MFCD29765749
InChI Key VWUCIBOKNZGWLX-UHFFFAOYSA-N
Synonym Imidazolium Bromide
PubChem CID 12203052
IUPAC Name 1H-imidazole hydrobromide
SMILES Br.N1C=CN=C1

Specifications

Melting Point 221°C
Color White-Yellow
Quantity 1 g
Formula Weight 148.99
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Imidazole Hydrobromide (Low water content)