missing translation for 'onlineSavingsMsg'
Learn More

Imidazole, >98.5% (Dry Basis), Ultrapure

CAS: 288-32-4 | C3H4N2 | 68.08 g/mol

Supplier:  Thermo Fisher Scientific J17525A1

 View more versions of this product

Catalog No. AAJ17525A1


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Imidazole
88°C to 92°C
256°C
Plastic bottle
C3H4N2
1 kg
103853
imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole
N1C=CN=C1
68.08
CHEBI:16069
≥98.5%
Flakes or Powder
288-32-4
1.03
145°C
Dry basis
MFCD00005183
UN3263
1.03
RAXXELZNTBOGNW-UHFFFAOYSA-N
1 H-imidazole
795
68.08
Ultrapure

Chemical Identifiers

288-32-4
68.08
RAXXELZNTBOGNW-UHFFFAOYSA-N
795
1 H-imidazole
C3H4N2
MFCD00005183
imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole
CHEBI:16069
N1C=CN=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335-H360D-H500

DOTInformation : Hazard Class: 8; Packaging Group: III

EINECSNumber : 206-019-2

RTECSNumber : NI3325000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

Promotions

Promotions

RUO – Research Use Only