missing translation for 'onlineSavingsMsg'
Learn More

Imidazole, >98.5% (Dry Basis), Ultrapure

Catalog No. AAJ17525A7
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ17525A7 5 kg Plastic bottle
AAJ1752522 100 g Plastic bottle
AAJ1752536 500 g Plastic bottle
AAJ17525A1 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAJ17525A7 Supplier Thermo Fisher Scientific Supplier No. J17525A7
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 288-32-4 | C3H4N2 | 68.08 g/mol

Chemical Identifiers

CAS 288-32-4
Molecular Formula C3H4N2
Molecular Weight (g/mol) 68.08
MDL Number MFCD00005183
InChI Key RAXXELZNTBOGNW-UHFFFAOYSA-N
Synonym imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole
PubChem CID 795
ChEBI CHEBI:16069
IUPAC Name 1 H-imidazole
SMILES N1C=CN=C1

Specifications

Melting Point 88°C to 92°C
Density 1.03
Boiling Point 256°C
Flash Point 145°C
Packaging Plastic bottle
Quantity 5 kg
UN Number UN3263
Beilstein 103853
Specific Gravity 1.03
Formula Weight 68.08
Percent Purity ≥98.5%
Grade Ultrapure
Physical Form Flakes or Powder
Assay Dry basis
Chemical Name or Material Imidazole
Show More Show Less
Hazard Category H302-H314-H335-H360D-H500
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 206-019-2
RTECSNumber NI3325000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only