missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Imidazole, 0.5M buffer soln., pH 7.0

Catalog No. AAJ62593AE
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62593AE 100 mL
AAJ62593AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62593AE Supplier Thermo Scientific Chemicals Supplier No. J62593AE

CAS: 288-32-4 | C3H4N2 | 68.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 288-32-4
Molecular Formula C3H4N2
Molecular Weight (g/mol) 68.079
MDL Number MFCD00005183
InChI Key RAXXELZNTBOGNW-UHFFFAOYSA-N
IUPAC Name 1H-imidazole
SMILES N1C=CN=C1

Specifications

Concentration 0.5M buffer soln.
pH 7.0
Physical Form Liquid
Quantity 100 mL
Recommended Storage Keep cold

RUO – Research Use Only