missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Imidazole, 0.2M buffer soln., pH 7.0, low endotoxin
$101.51 - $101.51
Specifications
| CAS | 288-32-4 |
|---|---|
| Color | Colorless |
| Concentration | 0.2M buffer soln. |
| pH | 7.0 |
| Physical Form | Liquid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ67055AE
|
Thermo Scientific™
J67055AE |
100 mL |
N/A
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| 288-32-4 | |
| 0.2M buffer soln. | |
| Liquid | |
| MFCD00005183 | |
| N1C=CN=C1 | |
| 68.079 |
| Colorless | |
| 7.0 | |
| C3H4N2 | |
| RAXXELZNTBOGNW-UHFFFAOYSA-N | |
| 1H-imidazole |