missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Imidazo[1,2-b]pyridazine, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 455660010
Specifications
| Imidazo[1,2-b]pyridazine | |
| Authentic | |
| Glass Bottle | |
| 1g | |
| Solubility in water: .. Other solubilities: soluble in dmf | |
| C1=CC2=NC=CN2N=C1 | |
| 119.12 | |
| 119.12 |
| 766-55-2 | |
| 97.5% min. (GC) | |
| C6H5N3 | |
| imidazo 1,2-b pyridazine, imidazol 1,2-b pyridazine, imdazo1,2-bpyridazine, pubchem11271, acmc-1bczx, imidazo 2,1-f pyridazine, 1,5-diazaindolizine, ksc258e5b, 1,3a,4-triazaindene | |
| VTVRXITWWZGKHV-UHFFFAOYSA-N | |
| imidazo[1,2-b]pyridazine | |
| 136599 | |
| 98% |
Chemical Identifiers
| 766-55-2 | |
| 119.12 | |
| imidazo 1,2-b pyridazine, imidazol 1,2-b pyridazine, imdazo1,2-bpyridazine, pubchem11271, acmc-1bczx, imidazo 2,1-f pyridazine, 1,5-diazaindolizine, ksc258e5b, 1,3a,4-triazaindene | |
| imidazo[1,2-b]pyridazine |
| C6H5N3 | |
| VTVRXITWWZGKHV-UHFFFAOYSA-N | |
| 136599 | |
| C1=CC2=NC=CN2N=C1 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
RUO â Research Use Only