missing translation for 'onlineSavingsMsg'
Learn More

Imidazo[1,2-a]pyridine 98.0+%, TCI America™

Catalog No. i09225g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I09225G Supplier TCI America Supplier No. I09225G
Only null left

Chemical Identifiers

CAS 274-76-0
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD00005553
InChI Key UTCSSFWDNNEEBH-UHFFFAOYSA-N
Synonym imidazo 1,2-a pyridine, unii-g18zbv2hxa, 1h-imidazo 1,2-a pyridine, g18zbv2hxa, pyrimidazole, 1-azaindolizine, 1,3a-diazaindene, pubchem17181, imidazo l,2-a pyridine
PubChem CID 78960
IUPAC Name imidazo[1,2-a]pyridine
SMILES C1=CN2C=CC=CC2=N1

Specifications

Boiling Point 98°C
Quantity 5 g
Formula Weight 118.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Imidazo[1,2-a]pyridine