accessibility menu, dialog, popup

UserName

CAS RN 274-76-0

IUPAC Name: imidazo[1,2-a]pyridine
Synonyms: imidazo[1,2-a]pyridine
Molecular Weight (g/mol): 118.14
Molecular Formula: C7H6N2
InChi Key: UTCSSFWDNNEEBH-UHFFFAOYSA-N
SMILES: C1=CN2C=CC=CC2=N1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)