missing translation for 'onlineSavingsMsg'
Learn More

Imidazo[1,2-a]pyridine, 99%, Thermo Scientific™

Chemical Identifiers

CAS 274-76-0
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD00005553
InChI Key UTCSSFWDNNEEBH-UHFFFAOYSA-N
Synonym imidazo 1,2-a pyridine, unii-g18zbv2hxa, 1h-imidazo 1,2-a pyridine, g18zbv2hxa, pyrimidazole, 1-azaindolizine, 1,3a-diazaindene, pubchem17181, imidazo l,2-a pyridine
PubChem CID 78960
IUPAC Name imidazo[1,2-a]pyridine
SMILES C1=CN2C=CC=CC2=N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC439940050
SDS View Documents
Thermo Scientific Chemicals
439940050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC439940250
SDS View Documents
Thermo Scientific Chemicals
439940250
25g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

274-76-0
118.14
UTCSSFWDNNEEBH-UHFFFAOYSA-N
78960
C1=CN2C=CC=CC2=N1
C7H6N2
MFCD00005553
imidazo 1,2-a pyridine, unii-g18zbv2hxa, 1h-imidazo 1,2-a pyridine, g18zbv2hxa, pyrimidazole, 1-azaindolizine, 1,3a-diazaindene, pubchem17181, imidazo l,2-a pyridine
imidazo[1,2-a]pyridine

Specifications

274-76-0
113°C
98.5% min. (GC)
C7H6N2
5g
UTCSSFWDNNEEBH-UHFFFAOYSA-N
imidazo[1,2-a]pyridine
78960
99%
97°C to 98°C
Authentic
Glass bottle
MFCD00005553
imidazo 1,2-a pyridine, unii-g18zbv2hxa, 1h-imidazo 1,2-a pyridine, g18zbv2hxa, pyrimidazole, 1-azaindolizine, 1,3a-diazaindene, pubchem17181, imidazo l,2-a pyridine
C1=CN2C=CC=CC2=N1
118.14
118.14
Imidazo[1, 2-a]pyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only