missing translation for 'onlineSavingsMsg'
Learn More

Imidazo[1,2-a]pyridine-6-carboxylic acid, 90%, Thermo Scientific™

Catalog No. CC33101DE
Change view
Click to view available options
Quantity:
250mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC33101DE 5g
CC33101CB 250mg
CC33101DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC33101DE Supplier Thermo Scientific Chemicals Supplier No. CC33101DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 139022-25-6
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.15
MDL Number MFCD07021498
InChI Key ONOJJCTXSDBVSP-UHFFFAOYSA-N
Synonym imidazo 1,2-a pyridine-6-carboxylic acid, h-imidazo 1,2-a pyridine-6-carboxylic acid, imidazo 1,2-a pyridine-6-carboxylicacid, 1h-imidazo 1,2-a pyridine-6-carboxylic acid, 4-hydroimidazo 1,2-a pyridine-6-carboxylic acid, pubchem23163, acmc-209cig, ksc173s9l, 6-carboxyimidazo 1,2-a pyridine, imidazol 1,2-a pyridine-6-carboxylic acid
PubChem CID 7075376
IUPAC Name imidazo[1,2-a]pyridine-6-carboxylic acid
SMILES OC(=O)C1=CN2C=CN=C2C=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5g
Formula Weight 162.15
Percent Purity 97%
Chemical Name or Material Imidazo[1, 2-a]pyridine-6-carboxylic acid
Hazard Category Warning