accessibility menu, dialog, popup

UserName

NNOOH

CAS RN 139022-25-6

IUPAC Name: imidazo[1,2-a]pyridine-6-carboxylic acid
Synonyms: imidazo[1,2-a]pyridine-6-carboxylic acid
Molecular Weight (g/mol): 162.15
Molecular Formula: C8H6N2O2
InChi Key: ONOJJCTXSDBVSP-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN2C=CN=C2C=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)