missing translation for 'onlineSavingsMsg'
Learn More

Imidazo[1,2-a]pyridine-6-carboxylic acid, 90%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC33101DA

Quantity 1g

 View more versions of this product

Catalog No. CC33101DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

139022-25-6
162.15
ONOJJCTXSDBVSP-UHFFFAOYSA-N
7075376
OC(=O)C1=CN2C=CN=C2C=C1
C8H6N2O2
MFCD07021498
imidazo 1,2-a pyridine-6-carboxylic acid, h-imidazo 1,2-a pyridine-6-carboxylic acid, imidazo 1,2-a pyridine-6-carboxylicacid, 1h-imidazo 1,2-a pyridine-6-carboxylic acid, 4-hydroimidazo 1,2-a pyridine-6-carboxylic acid, pubchem23163, acmc-209cig, ksc173s9l, 6-carboxyimidazo 1,2-a pyridine, imidazol 1,2-a pyridine-6-carboxylic acid
imidazo[1,2-a]pyridine-6-carboxylic acid

Specifications

Imidazo[1, 2-a]pyridine-6-carboxylic acid
97%
C8H6N2O2
1g
ONOJJCTXSDBVSP-UHFFFAOYSA-N
imidazo[1,2-a]pyridine-6-carboxylic acid
7075376
97%
139022-25-6
Amber Glass Bottle
MFCD07021498
imidazo 1,2-a pyridine-6-carboxylic acid, h-imidazo 1,2-a pyridine-6-carboxylic acid, imidazo 1,2-a pyridine-6-carboxylicacid, 1h-imidazo 1,2-a pyridine-6-carboxylic acid, 4-hydroimidazo 1,2-a pyridine-6-carboxylic acid, pubchem23163, acmc-209cig, ksc173s9l, 6-carboxyimidazo 1,2-a pyridine, imidazol 1,2-a pyridine-6-carboxylic acid
OC(=O)C1=CN2C=CN=C2C=C1
162.15
162.15
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Warning

SDS
Documents

Documents