Learn More
Imidazo[1,2-a]pyridine-6-carbonyl chloride hydrochloride , Tech., Thermo Scientific™
$70.17 - $70.17
Chemical Identifiers
| CAS | 859833-15-1 |
|---|---|
| Molecular Formula | C8H6Cl2N2O |
| Molecular Weight (g/mol) | 217.049 |
| MDL Number | MFCD07772809 |
| InChI Key | LHLAJCRPSZDOFF-UHFFFAOYSA-N |
| Synonym | imidazo 1,2-a pyridine-6-carbonyl chloride hydrochloride, imidazo 1,2-a pyridine-6-carbonylchloride, hydrochloride 1:1, 4-hydroimidazo 1,2-a pyridine-6-carbonyl chloride, chloride, imidazo 1,2-a pyridine-6-carbonyl chloride-hydrogen chloride 1/1 |
| PubChem CID | 18525720 |
| IUPAC Name | imidazo[1,2-a]pyridine-6-carbonyl chloride;hydrochloride |
| SMILES | C1=CC2=NC=CN2C=C1C(=O)Cl.Cl |
Chemical Identifiers
| 859833-15-1 | |
| 217.049 | |
| LHLAJCRPSZDOFF-UHFFFAOYSA-N | |
| 18525720 | |
| C1=CC2=NC=CN2C=C1C(=O)Cl.Cl |
| C8H6Cl2N2O | |
| MFCD07772809 | |
| imidazo 1,2-a pyridine-6-carbonyl chloride hydrochloride, imidazo 1,2-a pyridine-6-carbonylchloride, hydrochloride 1:1, 4-hydroimidazo 1,2-a pyridine-6-carbonyl chloride, chloride, imidazo 1,2-a pyridine-6-carbonyl chloride-hydrogen chloride 1/1 | |
| imidazo[1,2-a]pyridine-6-carbonyl chloride;hydrochloride |
Specifications
| 859833-15-1 | |
| 100.0 | |
| C8H6Cl2N2O | |
| MFCD07772809 | |
| imidazo 1,2-a pyridine-6-carbonyl chloride hydrochloride, imidazo 1,2-a pyridine-6-carbonylchloride, hydrochloride 1:1, 4-hydroimidazo 1,2-a pyridine-6-carbonyl chloride, chloride, imidazo 1,2-a pyridine-6-carbonyl chloride-hydrogen chloride 1/1 | |
| C1=CC2=NC=CN2C=C1C(=O)Cl.Cl | |
| 217.049 | |
| 217.05 | |
| Technical |
| 80.0 | |
| Amber Glass Bottle | |
| Yes | |
| 250 mg | |
| LHLAJCRPSZDOFF-UHFFFAOYSA-N | |
| imidazo[1,2-a]pyridine-6-carbonyl chloride;hydrochloride | |
| 18525720 | |
| Tech. | |
| Imidazo[1, 2-a]pyridine-6-carbonyl chloride hydrochloride |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger