accessibility menu, dialog, popup

UserName

CAS RN 859833-15-1

IUPAC Name: imidazo[1,2-a]pyridine-6-carbonyl chloride hydrochloride
Synonyms: imidazo[1,2-a]pyridine-6-carbonyl chloride hydrochloride
Molecular Weight (g/mol): 217.05
Molecular Formula: C8H6Cl2N2O
InChi Key: LHLAJCRPSZDOFF-UHFFFAOYSA-N
SMILES: Cl.ClC(=O)C1=CN2C=CN=C2C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)