missing translation for 'onlineSavingsMsg'
Learn More

Imidazo[1,2-a]pyridine-6-carbonyl chloride hydrochloride, 95%

Catalog No. aah3517403
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3517403 1 g
AAH35174MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3517403 Supplier Thermo Scientific Chemicals Supplier No. H3517403
Only null left

CAS: 859833-15-1 | C8H6Cl2N2O | 217.049 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 859833-15-1
Molecular Formula C8H6Cl2N2O
Molecular Weight (g/mol) 217.049
MDL Number MFCD07772809
InChI Key LHLAJCRPSZDOFF-UHFFFAOYSA-N
Synonym imidazo 1,2-a pyridine-6-carbonyl chloride hydrochloride, imidazo 1,2-a pyridine-6-carbonylchloride, hydrochloride 1:1, 4-hydroimidazo 1,2-a pyridine-6-carbonyl chloride, chloride, imidazo 1,2-a pyridine-6-carbonyl chloride-hydrogen chloride 1/1
PubChem CID 18525720
IUPAC Name imidazo[1,2-a]pyridine-6-carbonyl chloride;hydrochloride
SMILES C1=CC2=NC=CN2C=C1C(=O)Cl.Cl

Specifications

Melting Point 248°C to 250°C
Quantity 1 g
UN Number UN3261
Sensitivity Hygroscopic
Formula Weight 217.06
Percent Purity 95%
Chemical Name or Material Imidazo[1,2-a]pyridine-6-carbonyl chloride hydrochloride
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only