missing translation for 'onlineSavingsMsg'
Learn More

Imidazo[1,2-a]pyridine-6-carbaldehyde, ≥97%, Thermo Scientific™

Catalog No. CC33104CB
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC33104CB 250 mg Amber Glass Bottle
CC33104DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC33104CB Supplier Thermo Scientific Chemicals Supplier No. CC33104CB

Chemical Identifiers

CAS 116355-16-9
Molecular Formula C8H6N2O
Molecular Weight (g/mol) 146.149
MDL Number MFCD07772808
InChI Key VDWYLKPRUHCLAW-UHFFFAOYSA-N
Synonym imidazo 1,2-a pyridine-6-carbaldehyde, imidazo 1,2-a pyridine-6-carboxaldehyde, 6-formylimidazo 1,2-a pyridine, pubchem23883, acmc-20a31e, 6-imidazo 1,2-a pyridinecarbaldehyde, 6-imidazo 1,2-a-pyridinecarbaldehyde, h-imidazo 1,2-a pyridine-6-carbaldehyde, 4-hydroimidazo 1,2-a pyridine-6-carbaldehyde
PubChem CID 7164582
IUPAC Name imidazo[1,2-a]pyridine-6-carbaldehyde
SMILES C1=CC2=NC=CN2C=C1C=O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 146.15
Percent Purity 95%
Chemical Name or Material Imidazo[1, 2-a]pyridine-6-carbaldehyde
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.