accessibility menu, dialog, popup

UserName

NNO

CAS RN 116355-16-9

IUPAC Name: imidazo[1,2-a]pyridine-6-carbaldehyde
Synonyms: imidazo[1,2-a]pyridine-6-carbaldehyde
Molecular Weight (g/mol): 146.15
Molecular Formula: C8H6N2O
InChi Key: VDWYLKPRUHCLAW-UHFFFAOYSA-N
SMILES: O=CC1=CN2C=CN=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results