missing translation for 'onlineSavingsMsg'
Learn More

Imidazo[1,2-a]pyridine-3-carboxylic acid, 97%, Thermo Scientific™

Catalog No. p-3110986
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC25401CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC25401CB Supplier Thermo Scientific Chemicals Supplier No. CC25401CB

Chemical Identifiers

CAS 6200-60-8
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.15
MDL Number MFCD01720458
InChI Key DOGXPDFZEQXZDS-UHFFFAOYSA-N
Synonym imidazo 1,2-a pyridine-3-carboxylic acid, imidazo 1,2-a pyridine-3-carboxylicacid, pubchem19678, wln: t56 an dnj bvq, 3-carboxyimidazo 1,2-a pyridine, h-imidazo 1,2-a pyridine-3-carboxylic acid, 4-hydroimidazo 1,2-a pyridine-3-carboxylic acid, 1h-imidazo 1,2-a pyridine-3-carboxylic acid
PubChem CID 73142
IUPAC Name imidazo[1,2-a]pyridine-3-carboxylic acid
SMILES OC(=O)C1=CN=C2C=CC=CN12

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 162.15
Percent Purity 97%
Chemical Name or Material Imidazo[1, 2-a]pyridine-3-carboxylic acid
Hazard Category Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.