accessibility menu, dialog, popup

UserName

HOONN

CAS RN 6200-60-8

IUPAC Name: imidazo[1,2-a]pyridine-3-carboxylic acid
Synonyms: imidazo[1,2-a]pyridine-3-carboxylic acid
Molecular Weight (g/mol): 162.15
Molecular Formula: C8H6N2O2
InChi Key: DOGXPDFZEQXZDS-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=C2C=CC=CN12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results