missing translation for 'onlineSavingsMsg'
Learn More

Imidazo[1,2-a]pyridine-2-carboxylic acid monohydrate, 97%, Thermo Scientific™

Chemical Identifiers

CAS 64951-08-2
Molecular Formula C8H6N2O2·H2O
Molecular Weight (g/mol) 180.16
MDL Number MFCD17171427
InChI Key WQLJLPDGSLZYEP-UHFFFAOYSA-N
Synonym imidazo 1,2-a pyridine-2-carboxylic acid, imidazo 1,2-a pyridine-2-carboxylicacid, h-imidazo 1,2-a pyridine-2-carboxylic acid, 4-hydroimidazo 1,2-a pyridine-2-carboxylic acid, imidazo 1,2-a pyridine-2-carboxylic, 1h-imidazo 1,2-a pyridine-2-carboxylic acid, imidazo 1,2-a pyridine-2-carboxylic acid, technical grade
PubChem CID 2062168
IUPAC Name imidazo[1,2-a]pyridine-2-carboxylic acid
SMILES C1=CC2=NC(=CN2C=C1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC432780010
SDS View Documents
Thermo Scientific Chemicals
432780010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC432780050
SDS View Documents
Thermo Scientific Chemicals
432780050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

64951-08-2
180.16
WQLJLPDGSLZYEP-UHFFFAOYSA-N
2062168
C1=CC2=NC(=CN2C=C1)C(=O)O
C8H6N2O2·H2O
MFCD17171427
imidazo 1,2-a pyridine-2-carboxylic acid, imidazo 1,2-a pyridine-2-carboxylicacid, h-imidazo 1,2-a pyridine-2-carboxylic acid, 4-hydroimidazo 1,2-a pyridine-2-carboxylic acid, imidazo 1,2-a pyridine-2-carboxylic, 1h-imidazo 1,2-a pyridine-2-carboxylic acid, imidazo 1,2-a pyridine-2-carboxylic acid, technical grade
imidazo[1,2-a]pyridine-2-carboxylic acid

Specifications

64951-08-2
Authentic
Glass Bottle
MFCD17171427
imidazo 1,2-a pyridine-2-carboxylic acid, imidazo 1,2-a pyridine-2-carboxylicacid, h-imidazo 1,2-a pyridine-2-carboxylic acid, 4-hydroimidazo 1,2-a pyridine-2-carboxylic acid, imidazo 1,2-a pyridine-2-carboxylic, 1h-imidazo 1,2-a pyridine-2-carboxylic acid, imidazo 1,2-a pyridine-2-carboxylic acid, technical grade
C1=CC2=NC(=CN2C=C1)C(=O)O
180.16
180.16
Imidazo[1,2-a]pyridine-2-carboxylic acid monohydrate
51.20 to 55.47% C, 4.30 to 4.65% H, 14.96 to 16.17% N
96% min. (HPLC)
C8H6N2O2·H2O
1g
WQLJLPDGSLZYEP-UHFFFAOYSA-N
imidazo[1,2-a]pyridine-2-carboxylic acid
2062168
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only