missing translation for 'onlineSavingsMsg'
Learn More

Imidazo[1,2-a]pyridin-6-ylmethanol, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC33109DA

Quantity 1g

 View more versions of this product

Catalog No. CC33109DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

132213-07-1
148.165
imidazo 1,2-a pyridin-6-ylmethanol, 6-hydroxymethyl imidazo 1,2-a pyridine, imidazo 1,2-a pyridine-6-methanol, imidazo 1,2-a pyridin-6-yl-methanol, imidazo 1,2-a pyridin-6-yl methanol, acmc-209zmz, imidazo 1,2-a pyridin-6yl methanol, 6-hydroxymethyl imidazo 1,2-a pyridin, 4-hydroimidazo 1,2-a pyridin-6-ylmethan-1-ol
imidazo[1,2-a]pyridin-6-ylmethanol
C8H8N2O
IEOADZOTWIMSMC-UHFFFAOYSA-N
7060530
C1=CC2=NC=CN2C=C1CO

Specifications

Imidazo[1, 2-a]pyridin-6-ylmethanol
97%
C8H8N2O
imidazo 1,2-a pyridin-6-ylmethanol, 6-hydroxymethyl imidazo 1,2-a pyridine, imidazo 1,2-a pyridine-6-methanol, imidazo 1,2-a pyridin-6-yl-methanol, imidazo 1,2-a pyridin-6-yl methanol, acmc-209zmz, imidazo 1,2-a pyridin-6yl methanol, 6-hydroxymethyl imidazo 1,2-a pyridin, 4-hydroimidazo 1,2-a pyridin-6-ylmethan-1-ol
C1=CC2=NC=CN2C=C1CO
148.165
148.16
132213-07-1
Amber Glass Bottle
1g
IEOADZOTWIMSMC-UHFFFAOYSA-N
imidazo[1,2-a]pyridin-6-ylmethanol
7060530
97%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions