missing translation for 'onlineSavingsMsg'
Learn More

Imidazo[1,2-a]pyridin-2-ylmethanol, ≥90%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC31309DA

 View more versions of this product

Catalog No. CC31309DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

Imidazo[1, 2-a]pyridin-2-ylmethanol
96.5
97%
C8H8N2O
1g
ROPHYEIJSUUKEO-UHFFFAOYSA-N
{imidazo[1,2-a]pyridin-2-yl}methanol
2795540
97%
82090-52-6
100.0
Amber Glass Bottle
MFCD06797471
imidazo 1,2-a pyridin-2-ylmethanol, imidazo 1,2-a pyridine-2-methanol, h-imidazo 1,2-a pyridin-2-yl methanol, 2-hydroxymethyl imidazo 1,2-a pyridine, imidazo 1,2-a pyridin-2-yl methanol, imidazo 1,2-a-pyridin-2-ylmethanol, 4-hydroimidazo 1,2-a pyridin-2-ylmethan-1-ol, imidazo 1,2-a pyridine-2-ylmethanol
OCC1=CN2C=CC=CC2=N1
148.17
148.16

Chemical Identifiers

82090-52-6
148.17
ROPHYEIJSUUKEO-UHFFFAOYSA-N
2795540
OCC1=CN2C=CC=CC2=N1
C8H8N2O
MFCD06797471
imidazo 1,2-a pyridin-2-ylmethanol, imidazo 1,2-a pyridine-2-methanol, h-imidazo 1,2-a pyridin-2-yl methanol, 2-hydroxymethyl imidazo 1,2-a pyridine, imidazo 1,2-a pyridin-2-yl methanol, imidazo 1,2-a-pyridin-2-ylmethanol, 4-hydroimidazo 1,2-a pyridin-2-ylmethan-1-ol, imidazo 1,2-a pyridine-2-ylmethanol
{imidazo[1,2-a]pyridin-2-yl}methanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions