missing translation for 'onlineSavingsMsg'
Learn More

Imidazo[1,2-a]pyridin-2-ylmethanol, ≥90%, Thermo Scientific™

Catalog No. p-3110974
Change view
Click to view available options
Quantity:
250mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC31309CB 250mg
CC31309DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC31309CB Supplier Thermo Scientific Chemicals Supplier No. CC31309CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 82090-52-6
Molecular Formula C8H8N2O
Molecular Weight (g/mol) 148.17
MDL Number MFCD06797471
InChI Key ROPHYEIJSUUKEO-UHFFFAOYSA-N
Synonym imidazo 1,2-a pyridin-2-ylmethanol, imidazo 1,2-a pyridine-2-methanol, h-imidazo 1,2-a pyridin-2-yl methanol, 2-hydroxymethyl imidazo 1,2-a pyridine, imidazo 1,2-a pyridin-2-yl methanol, imidazo 1,2-a-pyridin-2-ylmethanol, 4-hydroimidazo 1,2-a pyridin-2-ylmethan-1-ol, imidazo 1,2-a pyridine-2-ylmethanol
PubChem CID 2795540
IUPAC Name imidazo[1,2-a]pyridin-2-ylmethanol
SMILES OCC1=CN2C=CC=CC2=N1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250mg
Formula Weight 148.16
Percent Purity 97%
Chemical Name or Material Imidazo[1, 2-a]pyridin-2-ylmethanol
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.