Learn More
Imetit dihydrobromide, 98%
An extremely potent, high affinity agonist at H3 and H4 receptors | CAS: 32385-58-3 | C6H12Br2N4S | 332.06 g/mol
Supplier: Thermo Scientific Chemicals J62346MC
Description
An extremely potent, high affinity agonist at H3 and H4 receptors
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Imetit dihydrobromide | |
| White | |
| MFCD00153816 | |
| imetit dihydrobromide, imetit hydrobromide, s-2-imidazol-4-yl ethyl isothiourea, s-2-1h-imidazol-4-yl ethyl isothiourea dihydrobromide, 2-1h-imidazol-4-yl ethyl carbamimidothioate dihydrobromide, s-2-imidazol-4-yl ethyl isothiourea dihydrobromide, imetit dihydrobromide hplc, s-2-imidazol-4-yl ethyl isothiourea 2hbr, s-2-imidazol-4-yl ethyl isothiourea dihydrochloride, 2-2-1h-imidazol-5-yl ethyl isothiourea dihydrobromide | |
| DOBOYMKCRRLTRF-UHFFFAOYSA-N | |
| 2-(1H-imidazol-5-yl)ethyl carbamimidothioate;dihydrobromide | |
| 11957573 | |
| 332.06 | |
| Powder |
| 32385-58-3 | |
| C6H12Br2N4S | |
| 100 mg | |
| Freely soluble in water,practically insoluble in acetone /n | |
| Br.Br.NC(=N)SCCC1=CN=CN1 | |
| 332.06 | |
| CHEBI:64151 | |
| 98% |
Chemical Identifiers
| 32385-58-3 | |
| 332.06 | |
| DOBOYMKCRRLTRF-UHFFFAOYSA-N | |
| 11957573 | |
| 2-(1H-imidazol-5-yl)ethyl carbamimidothioate;dihydrobromide |
| C6H12Br2N4S | |
| MFCD00153816 | |
| imetit dihydrobromide, imetit hydrobromide, s-2-imidazol-4-yl ethyl isothiourea, s-2-1h-imidazol-4-yl ethyl isothiourea dihydrobromide, 2-1h-imidazol-4-yl ethyl carbamimidothioate dihydrobromide, s-2-imidazol-4-yl ethyl isothiourea dihydrobromide, imetit dihydrobromide hplc, s-2-imidazol-4-yl ethyl isothiourea 2hbr, s-2-imidazol-4-yl ethyl isothiourea dihydrochloride, 2-2-1h-imidazol-5-yl ethyl isothiourea dihydrobromide | |
| CHEBI:64151 | |
| Br.Br.NC(=N)SCCC1=CN=CN1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only