missing translation for 'onlineSavingsMsg'
Learn More

Imatinib 98.0+%, TCI America™

Catalog No. p-7131648
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0906100MG 100 mg
I09061G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0906100MG Supplier TCI America Supplier No. I0906100MG

Chemical Identifiers

CAS 152459-95-5
Molecular Formula C29H31N7O
Molecular Weight (g/mol) 493.62
MDL Number MFCD05662257
InChI Key KTUFNOKKBVMGRW-UHFFFAOYSA-N
PubChem CID 5291
ChEBI CHEBI:45783
IUPAC Name N-(4-methyl-3-{[4-(pyridin-3-yl)pyrimidin-2-yl]amino}phenyl)-4-[(4-methylpiperazin-1-yl)methyl]benzamide
SMILES CN1CCN(CC2=CC=C(C=C2)C(=O)NC2=CC=C(C)C(NC3=NC=CC(=N3)C3=CC=CN=C3)=C2)CC1

Specifications

Melting Point 209°C
Quantity 100 mg
Formula Weight 493.62
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Imatinib
RTECSNumber CV5585673
TSCA No
Recommended Storage Freezer