missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iloperidone 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 124°C |
| Color | White |
| Quantity | 25 mg |
| Formula Weight | 426.49 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Iloperidone |
Safety and Handling
| RTECSNumber | KM5777850 |
| TSCA | No |
| Recommended Storage | Freezer (-20 °C) |