CAS RN 133454-47-4
IUPAC Name:
1-(4-{3-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]propoxy}-3-methoxyphenyl)ethan-1-one
Synonyms:
iloperidone zomaril
Molecular Weight (g/mol):
426.49
Molecular Formula:
C24H27FN2O4
InChi Key:
XMXHEBAFVSFQEX-UHFFFAOYSA-N
SMILES:
COC1=CC(=CC=C1OCCCN1CCC(CC1)C1=NOC2=CC(F)=CC=C12)C(C)=O
1
–
1
of
1
results
1
Iloperidone 98.0+%, TCI America™
CAS: 133454-47-4 Molecular Formula: C24H27FN2O4 Molecular Weight (g/mol): 426.49 MDL Number: MFCD00866688 InChI Key: XMXHEBAFVSFQEX-UHFFFAOYSA-N PubChem CID: 71360 ChEBI: CHEBI:65173 IUPAC Name: 1-(4-{3-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]propoxy}-3-methoxyphenyl)ethan-1-one SMILES: COC1=CC(=CC=C1OCCCN1CCC(CC1)C1=NOC2=CC(F)=CC=C12)C(C)=O