missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ IL4 (Interleukin 4); Recombinant Human Protein

Catalog No. CTP0043
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CTP0043 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CTP0043 Supplier Gibco™ Supplier No. CTP0043

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

IL4 (Interleukin 4); Recombinant Human Protein

Recombinant IL4 is a bioactive protein intended for use in cell culture applications. IL4 promotes the proliferation and differentiation of activated B cells. In addition, IL4 up-regulates class II MHC antigen expression and IgE receptors. ED50 = 0.05 to 0.4 ng⁄mL (Specific Activity: 2.0 x 10^7 to 2.5 x 10^6 units⁄mg). The biological activity is determined by measuring the dose-dependent proliferation of human TF-1 cells. A concentration range of 0.1 to 10.0 ng⁄mL is effective for most in vitro applications.

Specifications

For Use With (Application) Mammalian Cell Culture, Primary Cell Culture, Stem Cell Culture
Molecular Weight (g/mol) 14.9 kDa
Name IL4 (Interleukin 4) CTS™ Recombinant Human Protein
Purification Method Gel-purified
Quantity 1 mg
Storage Requirements 2°C to 8°C
Regulatory Status RUO
Packaging 1 Ea.
Gene Alias IL-4 (Interleukin 4)
Common Name IL4 (Interleukin 4) CTS™ Recombinant Human Protein
Product Type IL4 (Interleukin 4) CTS™ Recombinant Human Protein
Specific Reactivity 0.187 ng/mL
Species Human
Expression System E. coli
Protein Family Cytokines and receptors
Protein Subtype Interleukins
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.